accessibility menu, dialog, popup

UserName

NH2H2N

CAS RN 10217-52-4

IUPAC Name: hydrazine
Synonyms: hydrazine|diamine|nitrogen hydride|hydrazine base|hydrazyna|oxytreat 35|segidrin|rcra waste number U133
Molecular Weight (g/mol): 32.05
Molecular Formula: H4N2
InChi Key: OAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILES: NN
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)