accessibility menu, dialog, popup

UserName

NBrBr

CAS RN 101861-59-0

IUPAC Name: 3,5-dibromoquinoline
Synonyms: 3,5-dibromoquinoline
Molecular Weight (g/mol): 286.95
Molecular Formula: C9H5Br2N
InChi Key: IBADFXOMCWHDMS-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(Br)C=CC=C2N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)