accessibility menu, dialog, popup

UserName

CAS RN 10133-30-9

IUPAC Name: 1-benzothiophene-5-carbaldehyde
Synonyms: 1-benzothiophene-5-carbaldehyde
Molecular Weight (g/mol): 162.21
Molecular Formula: C9H6OS
InChi Key: QHHRWAPVYHRAJA-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results