accessibility menu, dialog, popup

UserName

CAS RN 1011-41-2

IUPAC Name: 2-(morpholin-4-yl)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-(morpholin-4-yl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 198.24
Molecular Formula: C8H10N2O2S
InChi Key: VDZWHWVAMDQEBT-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(S1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)