accessibility menu, dialog, popup

UserName

CAS RN 101079-29-2

H3CNCH3CH3H3CFPFFFFF

CAS RN 101079-29-2

IUPAC Name: hexadecyltrimethylazanium; hexafluoro-λ⁵-phosphanuide
Synonyms: cetyl-trimethyl-ammonium hexafluorophosphate
Molecular Weight (g/mol): 429.52
Molecular Formula: C19H42F6NP
InChi Key: XIHYIALOMDSESV-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCCCCCCCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results