accessibility menu, dialog, popup

UserName

NaAlOSOOOOSOOO

CAS RN 10102-71-3

IUPAC Name: aluminium(3+) sodium disulfate
Synonyms: sodium aluminium sulfate
Molecular Weight (g/mol): 242.08
Molecular Formula: AlNaO8S2
InChi Key: GJPYYNMJTJNYTO-UHFFFAOYSA-J
SMILES: [Na+].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)