accessibility menu, dialog, popup

UserName

NaNaOTeOOO

CAS RN 10101-83-4

IUPAC Name: disodium tellurate
Synonyms: sodium tellurate
Molecular Weight (g/mol): 237.58
Molecular Formula: Na2O4Te
InChi Key: XERQTZLDFHNZIC-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-][Te]([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results