accessibility menu, dialog, popup

UserName

CAS RN 10101-59-4

IUPAC Name: chromium(3+) tetrahydrate phosphate
Synonyms: chromium(3+) tetrahydrate phosphate
Molecular Weight (g/mol): 219.03
Molecular Formula: CrH8O8P
InChi Key: BVZHAJJRIRDSTG-UHFFFAOYSA-K
SMILES: O.O.O.O.[Cr+3].[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results