accessibility menu, dialog, popup

UserName

CAS RN 101-86-0

IUPAC Name: (2E)-2-(phenylmethylidene)octanal
Synonyms: hexyl cinnamic aldehyde 2-hexyl-3-phenyl-2-propenal α-hexylcinnamyl aldehyde α-hexyl-β-phenylacrolein hexyl cinnamaldehyde 2-(phenylmethylene)octanal 2-hexylcinnamaldehyde α-n-hexyl-β-phenylacrolein 2-benzylideneoctanal
Molecular Weight (g/mol): 216.32
Molecular Formula: C15H20O
InChi Key: GUUHFMWKWLOQMM-NTCAYCPXSA-N
SMILES: CCCCCC\C(C=O)=C/C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results