accessibility menu, dialog, popup

UserName

H3CO

CAS RN 1009-14-9

IUPAC Name: 1-phenylpentan-1-one
Synonyms: valerophenone pentanophenone
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: XKGLSKVNOSHTAD-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=CC=CC=C1
Boiling Point 
  • (4)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)