CAS RN 100641-16-5
IUPAC Name:
lutetium(3+) trinitrate
Synonyms:
lutetium(3+) trinitrate
Molecular Weight (g/mol):
360.98
Molecular Formula:
LuN3O9
InChi Key:
APRNQTOXCXOSHO-UHFFFAOYSA-N
SMILES:
[Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
1
of
1
results
1
Lutetium(III) nitrate hydrate, 99.9% (REO)
CAS: 100641-16-5 Molecular Formula: LuN3O9 Molecular Weight (g/mol): 360.98 MDL Number: MFCD00150421 InChI Key: APRNQTOXCXOSHO-UHFFFAOYSA-N Synonym: lutetium iii nitrate hydrate,lutetium nitrate hydrate,acmc-1buji,lu.3no3.h2o,lutetium nitrate-water 1/3/1,lutetium 3+ hydrate trinitrate PubChem CID: 16212852 SMILES: [Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O