accessibility menu, dialog, popup

UserName

IN

CAS RN 1006-47-9

IUPAC Name: 8-iodoquinoline
Synonyms: 8-iodoquinoline
Molecular Weight (g/mol): 255.06
Molecular Formula: C9H6IN
InChi Key: OBEGSKDGFBFMFJ-UHFFFAOYSA-N
SMILES: IC1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results