accessibility menu, dialog, popup

UserName

CAS RN 100516-54-9

IUPAC Name: benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Synonyms: benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol): 387.44
Molecular Formula: C24H21NO4
InChi Key: HECRUWTZAMPQOS-UHFFFAOYNA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results