accessibility menu, dialog, popup

UserName

CAS RN 10031-55-7

IUPAC Name: dieuropium(3+) octahydrate trisulfate
Synonyms: dieuropium(3+) octahydrate trisulfate
Molecular Weight (g/mol): 736.22
Molecular Formula: Eu2H16O20S3
InChi Key: PGDXJFDXKJLVTJ-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Eu+3].[Eu+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)