CAS RN 10031-27-3
IUPAC Name:
tetrabromo-λ⁴-tellane
Synonyms:
tellurium tetrabromide
Molecular Weight (g/mol):
447.22
Molecular Formula:
Br4Te
InChi Key:
PTYIPBNVDTYPIO-UHFFFAOYSA-N
SMILES:
Br[Te](Br)(Br)Br
1
–
1
of
1
results
1
Tellurium(IV) bromide, 99.9% (metals basis)
CAS: 10031-27-3 Molecular Formula: Br4Te Molecular Weight (g/mol): 447.22 MDL Number: MFCD00049575 InChI Key: PTYIPBNVDTYPIO-UHFFFAOYSA-N Synonym: tellurium tetrabromide,tellurium iv bromide,tellurium bromide tebr4 , t-4,unii-7a29efj1af,7a29efj1af,tetrabromo-,e4-tellane,tebr4,acmc-1bozh,tellurium iv bromide, ultra dry PubChem CID: 82311 IUPAC Name: tetrabromo-λ⁴-tellane SMILES: Br[Te](Br)(Br)Br