accessibility menu, dialog, popup

UserName

CAS RN 10029-09-1

IUPAC Name: 1-[4-(aminomethyl)cyclohexyl]methanamine
Synonyms: 1,4-cyclohexanedimethanamine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: OXIKYYJDTWKERT-UHFFFAOYSA-N
SMILES: NCC1CCC(CN)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results