accessibility menu, dialog, popup

UserName

CAS RN 1001907-57-8

IUPAC Name: (2-methyl-2H-indazol-6-yl)boronic acid
Synonyms: 2-methylindazol-6-ylboronic acid
Molecular Weight (g/mol): 175.98
Molecular Formula: C8H9BN2O2
InChi Key: JADBVQWCMHPPQA-UHFFFAOYSA-N
SMILES: CN1C=C2C=CC(=CC2=N1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results