accessibility menu, dialog, popup

UserName

CAS RN 1001264-89-6

IUPAC Name: 2-(4-chlorophenyl)-1-(4-{7-hydroxy-5-methyl-5H,6H,7H-cyclopenta[d]pyrimidin-4-yl}piperazin-1-yl)-3-[(propan-2-yl)amino]propan-1-one
Synonyms: 2-(4-chlorophenyl)-1-(4-{7-hydroxy-5-methyl-5H,6H,7H-cyclopenta[d]pyrimidin-4-yl}piperazin-1-yl)-3-(isopropylamino)propan-1-one
Molecular Weight (g/mol): 458
Molecular Formula: C24H32ClN5O2
InChi Key: GRZXWCHAXNAUHY-UHFFFAOYNA-N
SMILES: CC(C)NCC(C(=O)N1CCN(CC1)C1=NC=NC2=C1C(C)CC2O)C1=CC=C(Cl)C=C1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results