Please login to your online account to display your discounted pricing

Alfa Aesar™ Zwitterionic Acid Labile Surfactant II (ZALS II), Certified Mass Spec Grade

Chemical Identifiers

Chemical Name or Material Zwitterionic Acid Labile Surfactant II (ZALS II)
Purity Grade Certified Mass Spec Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67721LB0

 
alfa aesar™
J67721LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Zwitterionic Acid Labile Surfactant II (ZALS II)
Physical Form Lyophilized Powder
Purity Grade Certified Mass Spec Grade
Storage Note 1 -30° to -10°C
Color White
Odor Odorless
Quantity 5mg
Safety and Handling

-30° to -10°C