Please login to your online account to display your discounted pricing

Alfa Aesar™ Zwitterionic Acid Labile Surfactant I (ZALS I), Certified Mass Spec Grade

Chemical Identifiers

Chemical Name or Material Zwitterionic Acid Labile Surfactant I (ZALS I)
Purity Grade Certified Mass Spec Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67729LB0

  SDS
alfa aesar™
J67729LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Zwitterionic Acid Labile Surfactant I (ZALS I)
Physical Form Lyophilized Powder
Purity Grade Certified Mass Spec Grade
Color White
Odor Odorless
Storage Note 1 -30° to -10°C
Quantity 5mg
Safety and Handling

-30° to -10°C