Please login to your online account to display your discounted pricing

Zolantidine dimaleate, Tocris


Potent, centrally active H2 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS107010

 View more versions of this product

Catalog No. 107010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 613.68
Quantity 10mg
Synonym N-[3-[3-(1-Piperidinylmethyl)phenoxy]propyl]-2-benzothiazolamine dimaleate
CAS 104076-39-3
Assay Percent Range >98%
Chemical Name or Material Zolantidine dimaleate
Molecular Formula C22H27N3OS·2C4H4O4
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Zolantidine dimaleate
Assay Percent Range >98%
Molecular Formula C22H27N3OS·2C4H4O4
Formula Weight 613.68
Synonym N-[3-[3-(1-Piperidinylmethyl)phenoxy]propyl]-2-benzothiazolamine dimaleate
Accessories

Desiccate at +4°C