Please login to your online account to display your discounted pricing

Zolantidine dimaleate, Tocris

Potent, centrally active H2 antagonist

$195.49 - $816.90

Chemical Identifiers

Chemical Name or Material Zolantidine dimaleate
Assay Percent Range >98%
Molecular Formula C22H27N3OS·2C4H4O4
Formula Weight 613.68
Synonym N-[3-[3-(1-Piperidinylmethyl)phenoxy]propyl]-2-benzothiazolamine dimaleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

107050

 
r & d systems
RDS107050
50mg Each for $816.90

107010

 
r & d systems
RDS107010
10mg Each for $195.49
Description & Specifications

Specifications

Formula Weight 613.68
Quantity 50mg
CAS 104076-39-3
Chemical Name or Material Zolantidine dimaleate
Molecular Formula C22H27N3OS·2C4H4O4
Assay Percent Range >98%
Synonym N-[3-[3-(1-Piperidinylmethyl)phenoxy]propyl]-2-benzothiazolamine dimaleate
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C