Please login to your online account to display your discounted pricing

Zobell's Solution (ORP Standard, 220mV), LabChem™

$98.63 - $98.63

Chemical Identifiers

Chemical Name or Material Zobell's Solution
CAS 7732-18-5
CAS Max % 99
CAS 7447-40-7
CAS Max % 0.75
CAS 14459-95-1
CAS Max % 0.14
CAS 13746-66-2
CAS Max % 0.11
Name Note ORP Standard, 220mV
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC273101

 
lab chem
LC273101
Amber Poly 500mL Each for $98.63
Description & Specifications

Specifications

Formula Weight 329.26
Purity Grade Certified Grade
CAS Max % 0.14
CAS 7447-40-7
Solubility Soluble in water
CAS 14459-95-1
CAS 13746-66-2
Physical Form Liquid
Quantity 500mL
Density 1g/mL
CAS 7732-18-5
CAS Max % 0.11
CAS Max % 0.75
Linear Formula K3Fe(CN)66
Recommended Storage Refrigerate
Name Note ORP Standard, 220mV
Color clear, yellow solution
Concentration 220mV ± 10mV
Identification Passes Test
CAS Max % 99
Specific Gravity 1g/mL
Decomposition Information Potassium oxide; Carbon monoxide; Carbon dioxide
Chemical Name or Material Zobell's Solution
Packaging Amber Poly
Safety and Handling

Refrigerate