missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF747 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567036 Supplier Invitrogen™ Supplier No. PA567036

Rabbit Polyclonal Antibody

Immunogen sequence: PTCLPGPCAST GPAAPLGAAC Highest antigen sequence identity to the following orthologs - mouse 57%, rat 57%.

ZNF747 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.

Specifications

Antigen ZNF747
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF747
Gene Accession No. Q9BV97
Gene Alias KRAB domain-containing protein ZNF747; zinc finger protein 747; ZNF747
Gene Symbols ZNF747
Host Species Rabbit
Immunogen Recombinant Human ZNF747. Recombinant protein control fragment (Product #RP-107679).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less