Please login to your online account to display your discounted pricing

ZM 449829, Tocris

Potent, selective JAK3 inhibitor

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material ZM 449829
Assay Percent Range >98%
Molecular Formula C13H10O
Formula Weight 182.22
Synonym 1-(2-Naphthalenyl)-2-propen-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

136650

 
r & d systems
136650
50mg Each for $762.58

136610

 
r & d systems
136610
10mg Each for $181.19
Description & Specifications

Specifications

Formula Weight 182.22
Quantity 50mg
Chemical Name or Material ZM 449829
Molecular Formula C13H10O
Assay Percent Range >98%
CAS 4452-06-6
Synonym 1-(2-Naphthalenyl)-2-propen-1-one
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C