Please login to your online account to display your discounted pricing

ZM 39923 hydrochloride, Tocris

Potent, selective JAK3 inhibitor

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material ZM 39923 hydrochloride
Assay Percent Range >99%
Molecular Formula C23H25NO·HCl
Formula Weight 367.92
Synonym 3-benzylisopropylamino-1-naphthalen-2-yl-propan-1-one hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

136710

 
r & d systems
RDS136710
10mg Each for $221.01

136750

 
r & d systems
RDS136750
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 367.92
Quantity 10mg
Assay Percent Range >99%
CAS 58753-54-1
Chemical Name or Material ZM 39923 hydrochloride
Molecular Formula C23H25NO·HCl
Synonym 3-benzylisopropylamino-1-naphthalen-2-yl-propan-1-one hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C