Please login to your online account to display your discounted pricing

ZM 323881 hydrochloride, Tocris

Potent, selective inhibitor of VEGFR-2

$146.74 - $316.20

Chemical Identifiers

Chemical Name or Material ZM 323881 hydrochloride
Assay Percent Range >99%
Molecular Formula C22H18FN3O2·HCl
Formula Weight 411.86
Synonym 5-((7-Benzyloxyquinazolin-4-yl)amino)-4-fluoro-2-methylphenol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

24751

  SDS
r & d systems
24751
1mg Each for $146.74

247510

 
r & d systems
247510
10mg Each for $316.20
Description & Specifications

Specifications

Formula Weight 411.86
Quantity 1mg
Recommended Storage Desiccate at +4°C
Molecular Formula C22H18FN3O2·HCl
CAS 193000-39-4
Chemical Name or Material ZM 323881 hydrochloride
Synonym 5-((7-Benzyloxyquinazolin-4-yl)amino)-4-fluoro-2-methylphenol hydrochloride
Assay Percent Range >99%
Safety and Handling

Desiccate at +4°C