missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ZM-306416 hydrochloride, 98%, Thermo Scientific Chemicals
$94.21 - $555.55
Chemical Identifiers
CAS | 690206-97-4 |
---|---|
Molecular Formula | C16H13ClFN3O2 |
Molecular Weight (g/mol) | 333.747 |
MDL Number | MFCD08703133 |
InChI Key | YHUIUSRCUKUUQA-UHFFFAOYSA-N |
Synonym | n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine, anilinoquinazoline deriv. 4, d0c6ki, n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine, n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine |
PubChem CID | 5329006 |
IUPAC Name | N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine |
SMILES | COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67265MCR
|
Thermo Scientific Chemicals
J67265MCR |
1 mg |
Each for $94.21
|
|
|||||
AAJ67265MA
|
Thermo Scientific Chemicals
J67265MA |
10 mg |
Each for $555.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
690206-97-4 | |
333.747 | |
YHUIUSRCUKUUQA-UHFFFAOYSA-N | |
5329006 | |
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC |
C16H13ClFN3O2 | |
MFCD08703133 | |
n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine, anilinoquinazoline deriv. 4, d0c6ki, n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine, n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine | |
N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine |
Specifications
690206-97-4 | |
C16H13ClFN3O2 | |
1 mg | |
YHUIUSRCUKUUQA-UHFFFAOYSA-N | |
N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine | |
5329006 | |
98% | |
ZM-306416 hydrochloride |
White | |
MFCD08703133 | |
n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine, anilinoquinazoline deriv. 4, d0c6ki, n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine, n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine | |
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC | |
333.747 | |
370.21 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only