Please login to your online account to display your discounted pricing

ZK 93426 hydrochloride, Tocris

Potent, competitive benzodiazepine antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material ZK 93426 hydrochloride
Assay Percent Range >99%
Molecular Formula C18H20N2O3·HCl
Formula Weight 348.82
Synonym 4-Methyl-5-(1-methylethoxy)-9H-pyrido[3, 4-b]indole-3-carboxylic acid ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

199610

 
r & d systems
199610
10mg Each for $201.36

199650

 
r & d systems
199650
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 348.82
Quantity 10mg
CAS 1216792-30-1
Molecular Formula C18H20N2O3·HCl
Assay Percent Range >99%
Synonym 4-Methyl-5-(1-methylethoxy)-9H-pyrido[3, 4-b]indole-3-carboxylic acid ethyl ester hydrochloride
Chemical Name or Material ZK 93426 hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature