Please login to your online account to display your discounted pricing

ZK 93423 hydrochloride, Tocris

Potent benzodiazepine agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material ZK 93423 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H22N2O4·HCl
Formula Weight 426.9
Synonym 4-(Methoxymethyl)-6-(phenylmethoxy)-9H-pyrido[3, 4-b]indole-3-carboxylic acid ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

199410

 
r & d systems
199410
10mg Each for $225.77

199450

 
r & d systems
199450
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 426.9
Quantity 10mg
Chemical Name or Material ZK 93423 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H22N2O4·HCl
CAS 1216574-52-5
Synonym 4-(Methoxymethyl)-6-(phenylmethoxy)-9H-pyrido[3, 4-b]indole-3-carboxylic acid ethyl ester hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature