Please login to your online account to display your discounted pricing

ZK 756326, Tocris

Selective, non-peptide CCR8 agonist

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material ZK 756326
Assay Percent Range >99%
Molecular Formula C21H28N2O3·2HCl
Formula Weight 429.38
Synonym 2-[2-[4-[(3-phenoxyphenyl)methyl]-1-piperazinyl]ethoxy]ethanol dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

256550

 
r & d systems
256550
50mg Each for $812.12

256510

 
r & d systems
256510
10mg Each for $192.67
Description & Specifications

Specifications

Formula Weight 429.38
Quantity 50mg
Chemical Name or Material ZK 756326
Assay Percent Range >99%
Molecular Formula C21H28N2O3·2HCl
CAS 874911-96-3
Synonym 2-[2-[4-[(3-phenoxyphenyl)methyl]-1-piperazinyl]ethoxy]ethanol dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature