Please login to your online account to display your discounted pricing

Tocris™ ZK 164015

Potent estrogen receptor antagonist

$214.12 - $895.76

Specifications

Product Type ZK 164015
Inhibitors ZK 164015
Molecular Weight 513.73
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

218310

 
tocris™
218310
10mg Each for $214.12

218350

 
tocris™
218350
50mg Each for $895.76
Description & Specifications

Specifications

Product Type ZK 164015
Inhibitors ZK 164015
Molecular Weight 513.73
Purity >99

Potent estrogen receptor silent antagonist. Inhibits 17β-estradiol stimulation of luciferase activity (IC50 = 0.025μM); potently inhibits the growth of estrogen-sensitive human MCF-7 breast cancer cells in vitro (IC50 ∽ 1nM).

  • Chemical Name: 2-(4-Hydroxyphenyl)-3-methyl-1-[10-(pentylsulfonyl)decyl]-1H-indol-5-ol
  • Formula: C30H43NO4S
  • CAS Number: 177583-70-9
  • Solubility: Soluble to 25mM in ethanol and to 100mM in DMSO
  • Purity: >99