missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 222635000

 View more versions of this product

Catalog No. AC222635000


 

Specifications Description & Specifications

Specifications

Powder and/or Chunks
500g
degree of hydration: about 6 to 7
99.5%
N2O7Zr·xH2O
4% max.
ZrO(NO3)2·xH2O
MFCD00149896
231.23
White
99.4% min. calculated on the actual degree of hydration, 34.9 to 37.0% (ZrO2) calculated on the actual degree of hydration
99.50%
Zirconyl(IV) nitrate hydrate
Glass bottle
14985-18-3

Chemical Identifiers

14985-18-3
N2O7Zr·xH2O
231.23
Zirconyl(IV) nitrate hydrate, 99.5%
99.50%
ZrO(NO3)2·xH2O
MFCD00149896

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger