missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium wire, 1.14mm (0.045 in.) dia., 99.2% (metals basis excluding Hf), Hf 4.5% max

Manufacturer:  Alfa Aesar™ 14627G2

 View more versions of this product

Catalog No. AA14627G2



Specifications

Specifications

Zirconium wire
2m
7440-67-7
(metals basis excluding Hf), Hf 4.5% max.
MFCD00011303
Odorless
zirconium
23995
Wire
≈6.64g/m
1.14mm (0.045 in.) dia.
99.2%
Zr
QCWXUUIWCKQGHC-UHFFFAOYSA-N
[Zr]
91.224
CHEBI:33342

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
CHEBI:33342
[Zr]
Zr
MFCD00011303
23995
zirconium
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents