missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium wire, 0.5mm (0.020 in.) dia., annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max

Manufacturer:  Alfa Aesar™ 45056G5

 View more versions of this product

Catalog No. AA45056G5



Specifications

Specifications

Zirconium wire
5m
7440-67-7
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
Odorless
zirconium
23995
Wire
≈1.31g/m
0.5mm (0.020 in.) dia., Annealed
(metals basis excluding Hf) Hf 4.5% max
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
[Zr]
91.224
CHEBI:33342
99.2%

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents