Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm

Chemical Identifiers

Chemical Name or Material Zirconium titanium oxide
CAS 12036-70-3
Formula Weight 203.11
MDL Number MFCD00210648
Synonym Titanium zirconate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3949014

 
alfa aesar™
3949014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3949022

 
alfa aesar™
3949022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.11
Chemical Name or Material Zirconium titanium oxide
TSCA TSCA
Mesh Size −325 Mesh
Odor Odorless
Physical Form Powder
Assay 99.5%
Assay Percent Notes (metals basis excluding Hf), Hf <75ppm
MDL Number MFCD00210648
CAS 12036-70-3
Synonym Titanium zirconate
Quantity 25g