Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium telluride, 99.5% (metals basis excluding Hf), Hf 2-4%

Chemical Identifiers

Chemical Name or Material Zirconium telluride
CAS 32321-65-6
Formula Weight 346.42
MDL Number MFCD00145549
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3632106

 
alfa aesar™
3632106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3632114

 
alfa aesar™
3632114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 346.42
Chemical Name or Material Zirconium telluride
TSCA TSCA
Mesh Size −325 Mesh
Physical Form Powder
Assay 99.5%
Assay Percent Notes (metals basis excluding Hf), Hf 2 to 4%
MDL Number MFCD00145549
CAS 32321-65-6
Color Gray
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 6.42g/mL
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.