missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196471000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Zirconium standard solution | |
1 mg/ml Zr in 5% HF | |
Plastic bottle | |
Colorless | |
MFCD00011303 MFCD00148878 | |
[Zr] | |
91.22 | |
CHEBI:33342 | |
Liquid |
990 to 1010μg/mL of Zr(4+) in 5% HF | |
7440-67-7,7664-39-3,7732-18-5 | |
100 mL | |
Zr | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 | |
AAS |
RUO – Research Use Only