Please login to your online account to display your discounted pricing

Zirconium Standard, 1000mg/L in 2% HNO&subscript_3; (Assurance Grade), SPEX CertiPrep™

$81.31 - $81.31

Chemical Identifiers

Chemical Name or Material Zirconium Standard
CAS 7697-37-2
CAS Max % 2
CAS 13826-66-9
CAS Max % 0.25
CAS 7732-18-5
CAS Max % 97.75
Purity Grade Assurance Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLZR22M

 
spex certiprep llc
SPXPLZR22M
Triple-leached LDPE bottle 30mL Each for $81.31
Description & Specifications

Specifications

Quantity 30mL
Chemical Name or Material Zirconium Standard
CAS 7697-37-2
Health Hazard 1 May cause cancer.
Recommended Storage Ambient
CAS Max % 2
Name Note For AA and ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
CAS 13826-66-9
CAS Max % 0.25
DOT Information Nonhazardous
CAS 7732-18-5
CAS Max % 97.75
Concentration or Composition (by Analyte or Components) 1000mg/L
Packaging Triple-leached LDPE bottle
Solvent or Matrix 2% HNO3
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Physical Form Liquid
Purity Grade Assurance Grade

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

May cause cancer.

Ambient