Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis excluding Hf)

$623.80 - $623.80

Chemical Identifiers

Chemical Name or Material Zirconium sputtering target
CAS 7440-67-7
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41661KS

  SDS
alfa aesar™
41661KS
1 each Each for $623.80
Description & Specifications

Specifications

Chemical Name or Material Zirconium sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.5%
Assay Percent Notes (metals basis excluding Hf)
MDL Number MFCD00011303
CAS 7440-67-7
Odor Odorless
Quantity 1 each