Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis excluding Hf)

$559.01 - $559.01

Chemical Identifiers

Chemical Name or Material Zirconium sputtering target
CAS 7440-67-7
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41660KS

  SDS
alfa aesar™
41660KS
1 each Each for $559.01
Description & Specifications

Specifications

Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.5%
Chemical Name or Material Zirconium sputtering target
Odor Odorless
MDL Number MFCD00011303
CAS 7440-67-7
Quantity 1 each
TSCA TSCA
Assay Percent Notes (metals basis excluding Hf)