Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis excluding Hf)

Magnetron sputtering source

$415.86 - $415.86

Chemical Identifiers

Chemical Name or Material Zirconium sputtering target
CAS 7440-67-7
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41659KS

  SDS
alfa aesar™
41659KS
1 each Each for $415.86
Description & Specifications

Specifications

Chemical Name or Material Zirconium sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.5%
Assay Percent Notes (metals basis excluding Hf)
MDL Number MFCD00011303
CAS 7440-67-7
Odor Odorless
Quantity 1 each