missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium Sponge, 25.4mm (1.0 in.) and down, 99.2% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1419122

 View more versions of this product

Catalog No. AA1419122



Specifications

Specifications

Zirconium
7440-67-7
99.2% (Metals basis)
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
[Zr]
91.224
CHEBI:33342
Gray
100g
Gray sponge
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
Odorless
zirconium
23995
Sponge, 25.4mm (1.0 in.) and Down

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

H260-H252: In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities; may catch fire.

DOTInformation : DOT Class: 4.2, Packaging Group: III

EINECSNumber : 231-176-9

SDS Documents