Please login to your online account to display your discounted pricing

Zirconium Sponge, 25.4mm (1.0 in.) and down, 99.2% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Zirconium
CAS 7440-67-7
Physical Form Sponge, 25.4mm (1.0 in.) and Down
Assay Percent Range 99.2% (Metals basis)
MDL Number MFCD00011303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1419122

  SDS
alfa aesar
1419122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1419136

  SDS
alfa aesar
1419136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Sponge, 25.4mm (1.0 in.) and Down
Quantity 100g
Color Gray
DOT Information DOT Class: 4.2, Packaging Group: III
EINECS Number 231-176-9
Chemical Name or Material Zirconium
MDL Number MFCD00011303
Health Hazard 1 H260-H252: In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities; may catch fire.
CAS 7440-67-7
Odor Odorless
Appearance Gray sponge
Assay Percent Range 99.2% (Metals basis)
Safety and Handling

H260-H252: In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities; may catch fire.