Learn More
Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03632222
Description
Zirconium silicate is used for manufacturing refractory materials. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. zirconium silicate is as beads for milling and grinding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium silicide | |
12039-90-6 | |
100 g | |
H4Si2Zr | |
zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane | |
ZSONHIWWHDAZQO-UHFFFAOYSA-N | |
bis($l^{2}-silanylidene)zirconium | |
6336892 | |
99.5% | |
4.88 g/mL |
Gray | |
Powder | |
(metals basis excluding Hf), Hf 4% | |
MFCD00151547 | |
Insoluble in water. It is insoluble in acids,alkali and aqua regia. | |
[SiH2]=[Zr]=[SiH2] | |
151.43 | |
147.39 | |
Odorless |
Chemical Identifiers
12039-90-6 | |
151.43 | |
ZSONHIWWHDAZQO-UHFFFAOYSA-N | |
6336892 | |
[SiH2]=[Zr]=[SiH2] |
H4Si2Zr | |
MFCD00151547 | |
zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane | |
bis($l^{2}-silanylidene)zirconium |
Safety and Handling
EINECSNumber : 234-911-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only