missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium silicate, 98%, -325 mesh, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC258990010
Specifications
Zirconium silicate | |
10101-52-7 | |
0.7% to 2% Typically | |
O4SiZr | |
MFCD00085353 | |
a-pax 45m, hyacinth, standard sf 200, ultrox 500w, zircon, zircon zr sio4, zirconite, zirconium iv silicate, zirconium silicate, zircosil 15 | |
[O-][Si]([O-])([O-])[O-].[Zr+4] | |
183.305 | |
183.3 | |
63.0 to 66.0 % (ZrO2) 31.0 to 34.5 % (SiO2) | |
1.0% max. | |
325 mesh max. | |
1kg |
99%, -325 mesh | |
HfO2: 2% to 4% Typically | |
98% | |
ZrSiO4 | |
15, 10381 | |
GFQYVLUOOAAOGM-UHFFFAOYSA-N | |
zirconium(4+);silicate | |
61775 | |
Powder | |
0.05% to 0.1% Typically | |
Glass Bottle | |
Yellow to Orange |
Chemical Identifiers
10101-52-7 | |
183.305 | |
GFQYVLUOOAAOGM-UHFFFAOYSA-N | |
61775 | |
[O-][Si]([O-])([O-])[O-].[Zr+4] |
O4SiZr | |
MFCD00085353 | |
a-pax 45m, hyacinth, standard sf 200, ultrox 500w, zircon, zircon zr sio4, zirconite, zirconium iv silicate, zirconium silicate, zircosil 15 | |
zirconium(4+);silicate |
Safety and Handling
EINECSNumber : 233-252-7
TSCA : TSCA