Please login to your online account to display your discounted pricing

Zirconium silicate, 98%, -325 mesh, ACROS Organics™

$21.29 - $21.29

Chemical Identifiers

Chemical Name or Material Zirconium silicate
Name Note 99%, -325 mesh
CAS 10101-52-7
Assay Percent Range 98%
Molecular Formula SiO4Zr
Linear Formula ZrSiO4
Formula Weight 183.3
MDL Number MFCD00085353
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC258990010

  SDS
thermo fisher scientific
AC258990010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC258990100

  SDS
fisher scientific acros
258990100
Glass bottle 10g Each for $21.29

AC258991000

  SDS
thermo fisher scientific
258991000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 183.3
Quantity 1kg
Color Yellow to orange
TSCA TSCA
EINECS Number 233-252-7
Merck Index 15, 10381
Chemical Name or Material Zirconium silicate
Loss on Ignition 1.0% max.
Name Note 99%, -325 mesh
Packaging Glass bottle
Particle Size 325 mesh max.
Aluminum Oxide 0.7% to 2% Typically
Assay Percent Range 98%
Molecular Formula SiO4Zr
Linear Formula ZrSiO4
MDL Number MFCD00085353
CAS 10101-52-7
Additional Information HfO2: 2% to 4% Typically
Composition 63.0 to 66.0 % (ZrO2) 31.0 to 34.5 % (SiO2)
Fe2O3 0.05% to 0.1% Typically