Please login to your online account to display your discounted pricing

Zirconium silicate, 98%, -325 mesh, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  258991000

 View more versions of this product

Catalog No. AC258991000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 183.3
Quantity 100g
Color Yellow to orange
Merck Index 15, 10381
TSCA TSCA
EINECS Number 233-252-7
Particle Size 325 mesh max.
Fe2O3 0.05% to 0.1% Typically
Aluminum Oxide 0.7% to 2% Typically
Chemical Name or Material Zirconium silicate
Assay Percent Range 98%
Additional Information HfO2: 2% to 4% Typically
Packaging Glass bottle
Loss on Ignition 1.0% max.
Composition 63.0 to 66.0 % (ZrO2) 31.0 to 34.5 % (SiO2)
Name Note 99%, -325 mesh
Linear Formula ZrSiO4
MDL Number MFCD00085353
Molecular Formula SiO4Zr
CAS 10101-52-7

Specifications

Chemical Name or Material Zirconium silicate
Name Note 99%, -325 mesh
CAS 10101-52-7
Assay Percent Range 98%
Molecular Formula SiO4Zr
Linear Formula ZrSiO4
Formula Weight 183.3
MDL Number MFCD00085353