missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium rod, 99% (metals basis excluding Hf), Hf 4.5% max, Alfa Aesar™

Form: approx. 1.96g/cm, Temper: soft

Manufacturer:  Thermo Fisher Scientific 10443BW

 View more versions of this product

Catalog No. AA10443BW



Specifications

Specifications

Zirconium
Annealed
Gray to silver rod
Zr
and compounds
[Zr]
Odorless
23995
Rod, 6.2mm (0.24 in.) dia.
50cm
7440-67-7
99%
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
91.224
CHEBI:33342
Gray to Silver

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
and compounds
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

EINECSNumber : 231-176-9

Documents