missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium rod, 12.7mm (0.5 in.) dia., annealed, 99.2+% (metals basis excluding Hf), Hf 4.5% max

Manufacturer:  Alfa Aesar™ 10442BQ

 View more versions of this product

Catalog No. AA10442BQ



Specifications

Specifications

Zirconium rod
5cm
7440-67-7
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
Odorless
23995
Rod
≈8.24g/cm
12.7mm (0.5 in.) dia., Annealed, Temper: soft
(metals basis excluding Hf), Hf 4.5% max.
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
91.224
CHEBI:33342
≥99.2%

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents